siRNA Details
| Gene Symbol: Dctpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138892.1 | NP_620247.1 | 3 | 385 | ||
| GenBank mRNA | AF331839.1 | - | 3 | 359 | ||
| AY029335.1 | - | 3 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dctpp1| Gene Symbol: Dctpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138892.1 | NP_620247.1 | 3 | 385 | ||
| GenBank mRNA | AF331839.1 | - | 3 | 359 | ||
| AY029335.1 | - | 3 | 385 | |||