siRNA Details
| Gene Symbol: TFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206855.1 | NP_001193784.1 | 11 | 1257 | ||
| NM_003227.3 | NP_003218.2 | 14 | 1677 | |||
| XM_005250553.4 | XP_005250610.1 | 15 | 1983 | |||
| XM_017012573.1 | XP_016868062.1 | 15 | 1785 | |||
| GenBank mRNA | AF067864.1 | - | 14 | 1663 | ||
| AK000421.1 | - | 10 | 989 | |||
| BC142630.1 | - | 14 | 1663 | 40147690 | ||
| BC150192.1 | - | 11 | 1471 | |||
| BI917390.1 | - | 7 | 709 | |||

TFR2
Genomic Map