siRNA Details
| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138894.1 | NP_620249.1 | 7 | 712 | ||
| GenBank mRNA | AF374272.1 | - | 7 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grasp| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138894.1 | NP_620249.1 | 7 | 712 | ||
| GenBank mRNA | AF374272.1 | - | 7 | 712 | ||