siRNA Details
| Gene Symbol: Gls2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270786.1 | NP_001257715.1 | 7 | 1042 | ||
| NM_001270787.1 | NP_001257716.1 | 7 | 1288 | |||
| NM_138904.2 | NP_620259.2 | 6 | 737 | |||
| XM_006240728.2 | XP_006240790.1 | 7 | 1068 | |||
| XM_006240730.2 | XP_006240792.1 | 8 | 867 | |||
| GenBank mRNA | BC089776.1 | - | 7 | 881 | ||
| BC104712.1 | - | 7 | 786 | |||
| FQ211735.1 | - | 7 | 1288 | |||
| J05499.1 | - | 6 | 736 | |||

Gls2
Genomic Map