siRNA Details
| Gene Symbol: Sub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009618.1 | NP_001009618.1 | 4 | 330 | ||
| XM_006232043.2 | XP_006232105.1 | 4 | 297 | |||
| GenBank mRNA | BC088346.1 | - | 4 | 330 | 7368325 | |
| FQ211681.1 | - | 4 | 287 | |||
| FQ211748.1 | - | 4 | 287 | |||
| FQ228567.1 | - | 1 | 16 | |||
| FQ230867.1 | - | 4 | 260 | |||
| FQ232387.1 | - | 4 | 259 | |||
| K02816.1 | - | 3 | 185 | |||

Sub1
Genomic Map