siRNA Details
Gene Symbol: Adam6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138906.1 | NP_620261.1 | 1 | 937 | ||
GenBank mRNA | Y09111.1 | - | 1 | 937 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138906.1 | NP_620261.1 | 1 | 937 | ||
GenBank mRNA | Y09111.1 | - | 1 | 937 |