siRNA Details
| Gene Symbol: Acot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138907.2 | NP_620262.2 | 3 | 1183 | ||
| XM_006240320.2 | XP_006240382.1 | 3 | 1267 | |||
| GenBank mRNA | BC072540.1 | - | 3 | 1183 | 7104319 | |
| Y09333.1 | - | 3 | 1124 | |||

Acot2
Genomic Map