siRNA Details
| Gene Symbol: Dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138910.3 | NP_620265.1 | 3 | 621 | ||
| GenBank mRNA | BC061530.1 | - | 3 | 625 | 6917861 | |
| FQ222612.1 | - | 3 | 621 | |||
| FQ223332.1 | - | 3 | 630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dad1| Gene Symbol: Dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138910.3 | NP_620265.1 | 3 | 621 | ||
| GenBank mRNA | BC061530.1 | - | 3 | 625 | 6917861 | |
| FQ222612.1 | - | 3 | 621 | |||
| FQ223332.1 | - | 3 | 630 | |||