siRNA Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138911.3 | NP_620266.1 | 13, 14 | 1622 | ||
| GenBank mRNA | BC061529.1 | - | 13, 14 | 1623 | 6887497 | |
| FQ220758.1 | - | 10, 11 | 1144 | |||
| FQ227512.1 | - | 4, 5 | 373 | |||
| Y15068.1 | - | 12, 13 | 1557 | |||

Stip1
Genomic Map