siRNA Details
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138911.3 | NP_620266.1 | 2 | 258 | ||
GenBank mRNA | BC061529.1 | - | 2 | 259 | 6887497 | |
Y15068.1 | - | 1 | 193 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138911.3 | NP_620266.1 | 2 | 258 | ||
GenBank mRNA | BC061529.1 | - | 2 | 259 | 6887497 | |
Y15068.1 | - | 1 | 193 |