siRNA Details
| Gene Symbol: Ppp1r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138912.2 | NP_620267.2 | 2 | 492 | ||
| XM_006253215.2 | XP_006253277.1 | 2 | 1058 | |||
| XM_006253216.2 | XP_006253278.1 | 2 | 379 | |||
| GenBank mRNA | BC072514.1 | - | 2 | 492 | 7104413 | |
| Y18208.1 | - | 2 | 355 | |||
| Gene Symbol: RGD1562636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762746.1 | XP_008760968.1 | 1 | 224 | ||

Ppp1r3b
Genomic Map