siRNA Details
| Gene Symbol: Oas1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138913.1 | NP_620268.1 | 2 | 386 | ||
| XM_006249382.2 | XP_006249444.1 | 2 | 430 | |||
| GenBank mRNA | AY221507.1 | - | 2 | 402 | ||
| BC086979.1 | - | 2 | 373 | 7191865 | ||
| FQ231421.1 | - | 2 | 415 | |||
| FQ231433.1 | - | 2 | 391 | |||
| FQ232784.1 | - | 2 | 416 | |||
| Z18877.1 | - | 2 | 404 | |||

Oas1a
Genomic Map