siRNA Details
| Gene Symbol: Caly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190399.1 | NP_001177328.1 | 3 | 266 | ||
| NM_138915.1 | NP_620270.1 | 3 | 325 | |||
| GenBank mRNA | AF303658.1 | - | 3 | 325 | ||
| FQ213422.1 | - | 3 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caly| Gene Symbol: Caly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190399.1 | NP_001177328.1 | 3 | 266 | ||
| NM_138915.1 | NP_620270.1 | 3 | 325 | |||
| GenBank mRNA | AF303658.1 | - | 3 | 325 | ||
| FQ213422.1 | - | 3 | 240 | |||