siRNA Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013036.1 | NP_001013054.1 | 5 | 499 | ||
| XM_006251925.2 | XP_006251987.1 | 5 | 560 | |||
| XM_006251926.2 | XP_006251988.1 | 5 | 560 | |||
| XM_006251927.2 | XP_006251989.1 | 6 | 655 | |||
| GenBank mRNA | BC089848.1 | - | 5 | 499 | 7374684 | |
| FQ211507.1 | - | 6 | 595 | |||

Gmpr2
Genomic Map