siRNA Details
| Gene Symbol: Eml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138921.1 | NP_620276.1 | 6 | 577 | ||
| GenBank mRNA | AF335571.1 | - | 6 | 577 | ||
| BC062038.1 | - | 3 | 119 | 5598497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eml2| Gene Symbol: Eml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138921.1 | NP_620276.1 | 6 | 577 | ||
| GenBank mRNA | AF335571.1 | - | 6 | 577 | ||
| BC062038.1 | - | 3 | 119 | 5598497 | ||