siRNA Details
| Gene Symbol: Ppp1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138823.2 | NP_620178.1 | 615 | |||
| GenBank mRNA | BC092194.1 | - | 615 | 7313160 | ||
| S79213.1 | - | 630 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r2| Gene Symbol: Ppp1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138823.2 | NP_620178.1 | 615 | |||
| GenBank mRNA | BC092194.1 | - | 615 | 7313160 | ||
| S79213.1 | - | 630 | ||||