siRNA Details
| Gene Symbol: TGFB1I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042454.2 | NP_001035919.1 | 4 | 325 | ||
| NM_001164719.1 | NP_001158191.1 | 4 | 294 | |||
| NM_015927.4 | NP_057011.2 | 4 | 324 | |||
| GenBank mRNA | AB007836.1 | - | 3 | 289 | ||
| AF116343.1 | - | 3 | 240 | |||
| AK122975.1 | - | 2 | 1648 | |||
| AK313327.1 | - | 4 | 325 | |||
| BC001507.3 | - | 4 | 324 | 3030477 | ||
| BC001830.2 | - | 4 | 324 | 3030477 | ||
| BC017288.2 | - | 4 | 294 | 4901903 | ||
| BC032545.1 | - | 3 | 296 | 5535913 | ||
| DA097743.1 | - | 4 | 421 | |||

TGFB1I1
Genomic Map