siRNA Details
| Gene Symbol: Wbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138975.1 | NP_620431.1 | 1, 2 | 67 | ||
| XM_006247705.2 | XP_006247767.1 | 1, 2 | 147 | |||
| GenBank mRNA | AF499026.1 | - | 1, 2 | 67 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbp2| Gene Symbol: Wbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138975.1 | NP_620431.1 | 1, 2 | 67 | ||
| XM_006247705.2 | XP_006247767.1 | 1, 2 | 147 | |||
| GenBank mRNA | AF499026.1 | - | 1, 2 | 67 | ||