siRNA Details
| Gene Symbol: Acvr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139090.1 | NP_620790.1 | 2 | 391 | ||
| GenBank mRNA | U35025.1 | - | 2 | 276 | ||
| U69702.1 | - | 2 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acvr1c| Gene Symbol: Acvr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139090.1 | NP_620790.1 | 2 | 391 | ||
| GenBank mRNA | U35025.1 | - | 2 | 276 | ||
| U69702.1 | - | 2 | 391 | |||