siRNA Details
| Gene Symbol: Tmem37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139095.1 | NP_620795.1 | 2 | 64 | ||
| XM_008769424.1 | XP_008767646.1 | 2 | 424 | |||
| GenBank mRNA | AF361355.1 | - | 2 | 64 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem37| Gene Symbol: Tmem37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139095.1 | NP_620795.1 | 2 | 64 | ||
| XM_008769424.1 | XP_008767646.1 | 2 | 424 | |||
| GenBank mRNA | AF361355.1 | - | 2 | 64 | ||