siRNA Details
| Gene Symbol: Ddx46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139098.1 | NP_620798.1 | 11 | 1531 | ||
| XM_006253601.2 | XP_006253663.1 | 11 | 1561 | |||
| GenBank mRNA | BC107590.1 | - | 11 | 1397 | 7461248 | |
| U25746.1 | - | 11 | 1531 | |||

Ddx46
Genomic Map