siRNA Details
| Gene Symbol: Cd48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139103.1 | NP_620803.1 | 3 | 504 | ||
| GenBank mRNA | BC060542.1 | - | 3 | 541 | 6921213 | |
| FQ228817.1 | - | 3 | 558 | |||
| FQ234539.1 | - | 3 | 538 | |||
| X13016.1 | - | 3 | 504 | |||

Cd48
Genomic Map