siRNA Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139104.1 | NP_620804.1 | 10 | 1762 | ||
| GenBank mRNA | AF223678.1 | - | 10 | 1762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egfl7| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139104.1 | NP_620804.1 | 10 | 1762 | ||
| GenBank mRNA | AF223678.1 | - | 10 | 1762 | ||