siRNA Details
| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139108.2 | NP_620808.1 | 741 | |||
| GenBank mRNA | AF290194.1 | - | 746 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd5| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139108.2 | NP_620808.1 | 741 | |||
| GenBank mRNA | AF290194.1 | - | 746 | |||