siRNA Details
| Gene Symbol: Setd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113747.1 | NP_001107219.1 | 1483 | |||
| XM_006248028.2 | XP_006248090.1 | 5781 | ||||
| XM_008768559.1 | XP_008766781.1 | 7999 | ||||
| XM_008768560.1 | XP_008766782.1 | 4777 | ||||
| XM_008768561.1 | XP_008766783.1 | 4510 | ||||
| GenBank mRNA | AF388528.1 | - | 5, 6 | 856 | ||
| BC079469.1 | - | 1319 | ||||
| BC158669.1 | - | 1483 | 8370827 | |||

Setd4
Genomic Map