siRNA Details
| Gene Symbol: Lmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139112.2 | NP_620812.2 | 2 | 217 | ||
| XM_006229968.1 | XP_006230030.1 | 3 | 519 | |||
| GenBank mRNA | BF420017.1 | - | 2 | 217 | ||
| BF567497.1 | - | 1 | 29 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo1| Gene Symbol: Lmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139112.2 | NP_620812.2 | 2 | 217 | ||
| XM_006229968.1 | XP_006230030.1 | 3 | 519 | |||
| GenBank mRNA | BF420017.1 | - | 2 | 217 | ||
| BF567497.1 | - | 1 | 29 | |||