siRNA Details
| Gene Symbol: Coro6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139115.2 | NP_620815.2 | 7 | 907 | ||
| XM_006246885.2 | XP_006246947.1 | 8 | 1984 | |||
| XM_006246886.2 | XP_006246948.1 | 8 | 1986 | |||
| XM_006246887.2 | XP_006246949.1 | 8 | 1984 | |||
| XM_006246888.2 | XP_006246950.1 | 8 | 1986 | |||
| XM_006246889.2 | XP_006246951.1 | 7 | 1864 | |||
| XM_006246890.2 | XP_006246952.1 | 7 | 1864 | |||
| XM_006246891.2 | XP_006246953.1 | 8 | 1985 | |||
| GenBank mRNA | AF140359.1 | - | 8 | 1280 | ||
| BC080241.1 | - | 7 | 2446 | |||
| BC098656.1 | - | 7 | 907 | 7192854 | ||

Coro6
Genomic Map