siRNA Details
| Gene Symbol: Cdkn1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033757.1 | NP_001028929.1 | 3 | 1593 | ||
| NM_001033758.1 | NP_001028930.1 | 3 | 1557 | |||
| NM_182735.2 | NP_877399.2 | 4 | 1422 | |||
| XM_008760076.1 | XP_008758298.1 | 4 | 1505 | |||
| XM_008760077.1 | XP_008758299.1 | 4 | 1506 | |||
| XM_008760078.1 | XP_008758300.1 | 4 | 1465 | |||
| XM_008760079.1 | XP_008758301.1 | 4 | 1463 | |||
| GenBank mRNA | AY351678.1 | - | 3 | 1595 | ||
| AY351679.1 | - | 3 | 1559 | |||
| AY576608.1 | - | 4 | 1422 | |||
| AY586728.1 | - | 4 | 1387 | |||
| BC098646.1 | - | 4 | 1420 | 7109418 | ||
| CF110214.1 | - | 3 | 738 | |||

Cdkn1c
Genomic Map