siRNA Details
| Gene Symbol: Tubb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139254.2 | NP_640347.2 | 3 | 309 | ||
| GenBank mRNA | BC097281.1 | - | 3 | 289 | 7461390 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubb3| Gene Symbol: Tubb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139254.2 | NP_640347.2 | 3 | 309 | ||
| GenBank mRNA | BC097281.1 | - | 3 | 289 | 7461390 | |