siRNA Details
| Gene Symbol: Man2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139256.1 | NP_640349.1 | 8 | 1081 | ||
| GenBank mRNA | BC086989.1 | - | 8 | 962 | 7193864 | |
| M57547.1 | - | 8 | 1081 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man2c1| Gene Symbol: Man2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139256.1 | NP_640349.1 | 8 | 1081 | ||
| GenBank mRNA | BC086989.1 | - | 8 | 962 | 7193864 | |
| M57547.1 | - | 8 | 1081 | |||