siRNA Details
| Gene Symbol: Bmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139258.1 | NP_640351.1 | 3, 4 | 527 | ||
| XM_006234723.2 | XP_006234785.1 | 3, 4 | 565 | |||
| XM_006234724.2 | XP_006234786.1 | 3, 4 | 568 | |||
| GenBank mRNA | AF506761.1 | - | 3, 4 | 527 | ||

Bmf
Genomic Map