siRNA Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139259.2 | NP_640352.1 | 3 | 348 | ||
| GenBank mRNA | AF497263.1 | - | 3 | 352 | ||
| AF534395.1 | - | 2 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nradd| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139259.2 | NP_640352.1 | 3 | 348 | ||
| GenBank mRNA | AF497263.1 | - | 3 | 352 | ||
| AF534395.1 | - | 2 | 164 | |||