siRNA Details
| Gene Symbol: Gbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271453.1 | NP_001258382.1 | 1 | 1085 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbx1| Gene Symbol: Gbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271453.1 | NP_001258382.1 | 1 | 1085 | ||