siRNA Details
| Gene Symbol: Akap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037093.2 | NP_001032170.2 | 35, 36 | 9071 | ||
| XM_006236023.2 | XP_006236085.1 | 37, 38 | 9192 | |||
| XM_006236025.2 | XP_006236087.1 | 36, 37 | 9201 | |||
| XM_008762685.1 | XP_008760907.1 | 38, 39 | 9306 | |||
| XM_008762686.1 | XP_008760908.1 | 37, 38 | 9267 | |||
| GenBank mRNA | DQ228948.2 | - | 6, 7 | 1284 | ||

Akap9
Genomic Map