siRNA Details
| Gene Symbol: Akap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DQ228948.2 | - | 2 | 720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap9| Gene Symbol: Akap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DQ228948.2 | - | 2 | 720 | ||