siRNA Details
| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100823.1 | NP_001094293.1 | 19, 20 | 2878 | ||
| XM_006257442.2 | XP_006257504.1 | 19, 20 | 3065 | |||
| GenBank mRNA | AF130121.1 | - | 19, 20 | 2641 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr44| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100823.1 | NP_001094293.1 | 19, 20 | 2878 | ||
| XM_006257442.2 | XP_006257504.1 | 19, 20 | 3065 | |||
| GenBank mRNA | AF130121.1 | - | 19, 20 | 2641 | ||