siRNA Details
| Gene Symbol: Macrod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139337.1 | NP_647553.1 | 3 | 235 | ||
| XM_006230677.2 | XP_006230739.1 | 3 | 569 | |||
| GenBank mRNA | AF404762.1 | - | 3 | 235 | ||
| FQ216840.1 | - | 3 | 402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Macrod1| Gene Symbol: Macrod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139337.1 | NP_647553.1 | 3 | 235 | ||
| XM_006230677.2 | XP_006230739.1 | 3 | 569 | |||
| GenBank mRNA | AF404762.1 | - | 3 | 235 | ||
| FQ216840.1 | - | 3 | 402 | |||