siRNA Details
| Gene Symbol: Macrod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139337.1 | NP_647553.1 | 8, 9 | 691 | ||
| XM_006230677.2 | XP_006230739.1 | 8, 9 | 1025 | |||
| GenBank mRNA | AF404762.1 | - | 8, 9 | 691 | ||
| AF419856.1 | - | 8, 9 | 724 | |||
| FQ216840.1 | - | 8, 9 | 859 | |||

Macrod1
Genomic Map