siRNA Details
| Gene Symbol: Slc34a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139338.2 | NP_647554.1 | 10 | 1110 | ||
| XM_006233558.2 | XP_006233620.1 | 11 | 1323 | |||
| XM_006233559.2 | XP_006233621.1 | 11 | 1439 | |||
| XM_006233560.2 | XP_006233622.1 | 11 | 1300 | |||
| XM_006233561.2 | XP_006233623.1 | 10 | 1213 | |||
| XM_006233562.2 | XP_006233624.1 | 10 | 1329 | |||
| XM_006233564.2 | XP_006233626.1 | 10 | 1193 | |||
| XM_006233565.2 | XP_006233627.1 | 11 | 1299 | |||
| XM_006233566.2 | XP_006233628.1 | 10 | 1305 | |||
| XM_006233567.2 | XP_006233629.1 | 10 | 1181 | |||
| XM_008761577.1 | XP_008759799.1 | 11 | 1388 | |||
| GenBank mRNA | AB077042.1 | - | 10 | 1115 | ||

Slc34a3
Genomic Map