siRNA Details
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139339.1 | NP_647555.1 | 6 | 676 | ||
GenBank mRNA | AF512430.1 | - | 6 | 676 | ||
BC166414.1 | - | 6 | 610 | 9027758 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139339.1 | NP_647555.1 | 6 | 676 | ||
GenBank mRNA | AF512430.1 | - | 6 | 676 | ||
BC166414.1 | - | 6 | 610 | 9027758 |