siRNA Details
| Gene Symbol: Plekha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763357.1 | XP_008761579.1 | 8 | 778 | ||
| XM_008763358.1 | XP_008761580.1 | 8 | 778 | |||
| XM_008763359.1 | XP_008761581.1 | 8 | 778 | |||
| XM_008763360.1 | XP_008761582.1 | 8 | 778 | |||
| XM_008763361.1 | XP_008761583.1 | 6 | 1469 | |||
| XM_008763362.1 | XP_008761584.1 | 8 | 778 | |||
| XM_008763364.1 | XP_008761586.1 | 8 | 778 | |||
| GenBank mRNA | AF468695.1 | - | 6 | 527 | ||

Plekha5
Genomic Map