siRNA Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139342.1 | NP_647558.1 | 11 | 1744 | ||
| GenBank mRNA | AF036537.2 | - | 11 | 1744 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ripk3| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139342.1 | NP_647558.1 | 11 | 1744 | ||
| GenBank mRNA | AF036537.2 | - | 11 | 1744 | ||