siRNA Details
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139342.1 | NP_647558.1 | 8 | 1236 | ||
XM_006251928.2 | XP_006251990.1 | 7 | 2088 | |||
XM_008770661.1 | XP_008768883.1 | 6 | 1049 | |||
XM_008770662.1 | XP_008768884.1 | 7 | 1420 | |||
GenBank mRNA | AF036537.2 | - | 8 | 1236 | ||
BC158580.1 | - | 6 | 803 | 7378607 |
