siRNA Details
| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144738.2 | NP_653339.2 | 25 | 3013 | ||
| GenBank mRNA | AY100477.1 | - | 7 | 815 | ||
| BC083547.1 | - | 8 | 819 | 7132749 | ||
| BC091230.1 | - | 25 | 3013 | 7321805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnpla7| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144738.2 | NP_653339.2 | 25 | 3013 | ||
| GenBank mRNA | AY100477.1 | - | 7 | 815 | ||
| BC083547.1 | - | 8 | 819 | 7132749 | ||
| BC091230.1 | - | 25 | 3013 | 7321805 | ||