siRNA Details
| Gene Symbol: Fmo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144739.1 | NP_653340.1 | 9 | 1823 | ||
| GenBank mRNA | AF458413.1 | - | 9 | 1823 | ||
| BC070883.1 | - | 9 | 1551 | 7096939 | ||
| FQ218797.1 | - | 422 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmo5| Gene Symbol: Fmo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144739.1 | NP_653340.1 | 9 | 1823 | ||
| GenBank mRNA | AF458413.1 | - | 9 | 1823 | ||
| BC070883.1 | - | 9 | 1551 | 7096939 | ||
| FQ218797.1 | - | 422 | ||||