siRNA Details
| Gene Symbol: Adipoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144744.3 | NP_653345.1 | 3 | 778 | ||
| GenBank mRNA | AY033885.1 | - | 3 | 725 | ||
| BC092565.1 | - | 2 | 714 | 7308618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adipoq| Gene Symbol: Adipoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144744.3 | NP_653345.1 | 3 | 778 | ||
| GenBank mRNA | AY033885.1 | - | 3 | 725 | ||
| BC092565.1 | - | 2 | 714 | 7308618 | ||