siRNA Details
| Gene Symbol: Slc45a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144747.3 | NP_653348.3 | 3 | 528 | ||
| GenBank mRNA | AB075229.1 | - | 2 | 437 | ||
| BC093599.1 | - | 3 | 528 | 7315729 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc45a1| Gene Symbol: Slc45a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144747.3 | NP_653348.3 | 3 | 528 | ||
| GenBank mRNA | AB075229.1 | - | 2 | 437 | ||
| BC093599.1 | - | 3 | 528 | 7315729 | ||