siRNA Details
| Gene Symbol: Acsm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144748.1 | NP_653349.1 | 9 | 1226 | ||
| XM_008759677.1 | XP_008757899.1 | 9 | 1211 | |||
| GenBank mRNA | AF062389.1 | - | 9 | 1226 | ||
| BC078721.1 | - | 9 | 1203 | 7110246 | ||

Acsm2a
Genomic Map