siRNA Details
| Gene Symbol: Zfp382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144749.1 | NP_653350.1 | 1026 | |||
| GenBank mRNA | BC127483.1 | - | 439 | 8365988 | ||
| U56732.1 | - | 1026 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp382| Gene Symbol: Zfp382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144749.1 | NP_653350.1 | 1026 | |||
| GenBank mRNA | BC127483.1 | - | 439 | 8365988 | ||
| U56732.1 | - | 1026 | ||||