siRNA Details
| Gene Symbol: Aspg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144750.2 | NP_653351.1 | 14 | 1667 | ||
| GenBank mRNA | AB009372.1 | - | 14 | 1668 | ||
| BC098655.1 | - | 14 | 1637 | 7373402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aspg| Gene Symbol: Aspg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144750.2 | NP_653351.1 | 14 | 1667 | ||
| GenBank mRNA | AB009372.1 | - | 14 | 1668 | ||
| BC098655.1 | - | 14 | 1637 | 7373402 | ||